Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053748.1 | NP_446200.1 | 4 - 5 | 587 | 108 | |
XM_006230633.2 | XP_006230695.1 | 5 - 6 | 774 | 108 | ||
GenBank mRNA | BC107673.1 | - | 5 - 6 | 598 | 7938129 | 108 |
D89340.2 | - | 4 - 5 | 587 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |