Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139333.3 | NP_647549.2 | 15 - 16 | 1606 | 130 | |
XM_006231014.2 | XP_006231076.1 | 15 - 16 | 1697 | 130 | ||
GenBank mRNA | AB020022.2 | - | 15 - 16 | 1652 | 130 | |
BC107669.1 | - | 15 - 16 | 1606 | 7937645 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |