Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011940.1 | NP_001011940.1 | 3 - 4 | 398 | 72 | |
XM_006231029.2 | XP_006231091.1 | 3 - 4 | 435 | 72 | ||
GenBank mRNA | BC079048.1 | - | 3 - 4 | 398 | 7115295 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |