Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728854.4 | XP_002728900.1 | 7 - 8 | 1198 | 95 | |
XM_006230930.2 | XP_006230992.1 | 7 - 8 | 1259 | 95 | ||
XM_008760191.1 | XP_008758413.1 | 5 - 6 | 617 | 95 | ||
XM_008760192.1 | XP_008758414.1 | 5 - 6 | 571 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |