Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fra10ac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014246.1 | NP_001014268.1 | 3 - 4 | 306 | 160 | |
XM_008760371.1 | XP_008758593.1 | 3 - 4 | 1295 | 160 | ||
XM_008760372.1 | XP_008758594.1 | 3 - 4 | 1295 | 160 | ||
GenBank mRNA | BC090036.1 | - | 3 - 4 | 306 | 7368097 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |