Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013162.1 | NP_037294.1 | 2 - 3 | 162 | 92 | |
GenBank mRNA | BC167099.1 | - | 2 - 3 | 163 | 7369314 | 92 |
CO561550.1 | - | 2 - 3 | 162 | 92 | ||
FQ218638.1 | - | 2 - 3 | 178 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |