Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012739.1 | NP_001012757.1 | 1 - 2 | 56 | 70 | |
GenBank mRNA | BC091402.1 | - | 1 - 2 | 56 | 7324328 | 70 |
FQ211511.1 | - | 1 - 2 | 46 | 70 | ||
FQ214844.1 | - | 1 - 2 | 37 | 70 | ||
FQ223418.1 | - | 1 - 2 | 46 | 70 | ||
FQ231235.1 | - | 1 - 2 | 70 | |||
X62671.1 | - | 1 - 2 | 46 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |