Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ust5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760213.1 | XP_008758435.1 | 3 - 4 | 1478 | 99 | |
GenBank mRNA | BC081730.1 | - | 3 - 4 | 1184 | 99 | |
BC098905.1 | - | 3 - 4 | 841 | 7325342 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |