Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108513.1 | NP_001101983.1 | 7 - 8 | 1000 | 57 | |
XM_006230833.1 | XP_006230895.1 | 7 - 8 | 967 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |