Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031124.1 | NP_112386.1 | 4 - 5 | 370 | 72 | |
XM_006231097.2 | XP_006231159.1 | 4 - 5 | 454 | 72 | ||
XM_008760262.1 | XP_008758484.1 | 4 - 5 | 454 | 72 | ||
XM_008760263.1 | XP_008758485.1 | 4 - 5 | 454 | 72 | ||
XM_008760264.1 | XP_008758486.1 | 4 - 5 | 454 | 72 | ||
XM_008760265.1 | XP_008758487.1 | 4 - 5 | 454 | 72 | ||
XM_008760266.1 | XP_008758488.1 | 4 - 5 | 454 | 72 | ||
XM_008760267.1 | XP_008758489.1 | 4 - 5 | 454 | 72 | ||
GenBank mRNA | BC168246.1 | - | 4 - 5 | 324 | 5600098 | 72 |
L20820.1 | - | 4 - 5 | 370 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |