Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107587.1 | NP_001101057.1 | 1 - 2 | 363 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |