Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029927.1 | NP_001025098.1 | 4 - 5 | 433 | 94 | |
XM_006236511.2 | XP_006236573.2 | 4 - 5 | 909 | 94 | ||
XM_008762915.1 | XP_008761137.1 | 6 - 7 | 1218 | 94 | ||
GenBank mRNA | BC097471.1 | - | 4 - 5 | 433 | 7457768 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |