Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024797.1 | NP_001019968.1 | 2 - 3 | 765 | 66 | |
XM_006232735.2 | XP_006232797.1 | 2 - 3 | 756 | 66 | ||
XM_006232736.2 | XP_006232798.1 | 2 - 3 | 744 | 66 | ||
XM_006232737.2 | XP_006232799.1 | 2 - 3 | 744 | 66 | ||
XM_006232738.2 | XP_006232800.1 | 2 - 3 | 744 | 66 | ||
XM_006232741.2 | XP_006232803.1 | 2 - 3 | 744 | 66 | ||
XM_008761201.1 | XP_008759423.1 | 3 - 4 | 770 | 66 | ||
GenBank mRNA | AB195689.1 | - | 1 - 2 | 599 | 66 | |
AY724781.1 | - | 2 - 3 | 765 | 66 | ||
AY724782.1 | - | 2 - 3 | 765 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |