Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpini1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053779.1 | NP_446231.1 | 1 - 2 | 91 | 109 | |
GenBank mRNA | AF193014.1 | - | 1 - 2 | 91 | 109 | |
AF193015.1 | - | 1 - 2 | 97 | 109 | ||
BC061536.1 | - | 1 - 2 | 105 | 6920827 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |