Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025709.1 | NP_001020880.1 | 2 - 3 | 209 | 78 | |
GenBank mRNA | BC099162.1 | - | 2 - 3 | 209 | 7381935 | 78 |
FQ211568.1 | - | 318 | 78 | |||
FQ229403.1 | - | 2 - 3 | 194 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |