Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpgm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199382.1 | NP_955414.1 | 1 - 2 | 69 | 59 | |
GenBank mRNA | BC062240.1 | - | 1 - 2 | 69 | 6887677 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |