Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127497.1 | NP_001120969.1 | 12 - 13 | 929 | 126 | |
XM_006236363.2 | XP_006236425.1 | 12 - 13 | 1231 | 126 | ||
XM_008762821.1 | XP_008761043.1 | 12 - 13 | 1326 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |