Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127497.1 | NP_001120969.1 | 13 - 14 | 1028 | 67 | |
XM_006236363.2 | XP_006236425.1 | 13 - 14 | 1330 | 67 | ||
XM_008762821.1 | XP_008761043.1 | 13 - 14 | 1425 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |