Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100886.1 | NP_001094356.1 | 6 - 7 | 944 | 64 | |
XM_006240868.2 | XP_006240930.1 | 6 - 7 | 942 | 64 | ||
XM_006240869.2 | XP_006240931.1 | 6 - 7 | 941 | 64 | ||
XM_006240870.2 | XP_006240932.1 | 6 - 7 | 942 | 64 | ||
GenBank mRNA | BC100259.1 | - | 6 - 7 | 889 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |