Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241093.2 | XP_006241155.1 | 1 - 2 | 242 | 103 | |
XM_008765184.1 | XP_008763406.1 | 1 - 2 | 244 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |