Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240902.2 | XP_006240964.1 | 1 - 2 | 379 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |