Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109591.1 | NP_001103061.1 | 2 - 3 | 182 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |