Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008525.1 | NP_001008525.1 | 5 - 6 | 431 | 84 | |
XM_006240972.2 | XP_006241034.1 | 6 - 7 | 678 | 84 | ||
GenBank mRNA | BC086574.1 | - | 5 - 6 | 431 | 7306868 | 84 |
FQ232492.1 | - | 5 - 6 | 440 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |