Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080897.1 | NP_543173.1 | 3 - 4 | 282 | 100 | |
GenBank mRNA | AF454391.1 | - | 3 - 4 | 282 | 100 | |
FQ218385.1 | - | 3 - 4 | 286 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Rattus norvegicus Probe spans exons |