Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034124.1 | NP_001029296.1 | 4 - 5 | 361 | 83 | |
GenBank mRNA | BC100616.1 | - | 4 - 5 | 361 | 7933452 | 83 |
Gene Symbol: LOC102553715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246561.2 | XP_006246623.1 | 439 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |