Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034112.1 | NP_001029284.1 | 3 - 4 | 333 | 87 | |
XM_006247362.2 | XP_006247424.1 | 3 - 4 | 231 | 87 | ||
GenBank mRNA | BC128702.1 | - | 3 - 4 | 367 | 7323425 | 87 |
CO560831.1 | - | 3 - 4 | 333 | 87 | ||
DQ350891.1 | - | 3 - 4 | 170 | 87 | ||
DQ350892.1 | - | 3 - 4 | 170 | 87 | ||
DQ350893.1 | - | 3 - 4 | 170 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |