Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022404.2 | NP_071799.2 | 2 - 3 | 492 | 95 | |
GenBank mRNA | BC097319.1 | - | 2 - 3 | 170 | 7455095 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |