Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108365.1 | NP_001101835.1 | 4 - 5 | 1108 | 108 | |
XM_006251340.2 | XP_006251402.1 | 3 - 4 | 1143 | 108 | ||
XM_008770332.1 | XP_008768554.1 | 2 - 3 | 1469 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |