Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013193.1 | NP_037325.1 | 1 - 2 | 512 | 71 | |
XM_006251188.2 | XP_006251250.1 | 1 - 2 | 562 | 71 | ||
XM_006251191.2 | XP_006251253.1 | 1 - 2 | 562 | 71 | ||
GenBank mRNA | CB578019.1 | - | 1 - 2 | 512 | 71 | |
U61772.1 | - | 1 - 2 | 106 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |