Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013193.1 | NP_037325.1 | 14 - 15 | 1967 | 70 | |
XM_006251188.2 | XP_006251250.1 | 14 - 15 | 2017 | 70 | ||
GenBank mRNA | CR467414.1 | - | 1 - 2 | 70 | ||
U61772.1 | - | 14 - 15 | 1561 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |