Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110490.2 | NP_001103960.1 | 7 - 8 | 807 | 79 | |
NM_001277158.1 | NP_001264087.1 | 8 - 9 | 822 | 79 | ||
NM_001277159.1 | NP_001264088.1 | 8 - 9 | 812 | 79 | ||
NM_001277165.1 | NP_001264094.1 | 6 - 7 | 938 | 79 | ||
NM_001277166.1 | NP_001264095.1 | 7 - 8 | 797 | 79 | ||
XM_006252637.2 | XP_006252699.1 | 6 - 7 | 994 | 79 | ||
XM_006252640.2 | XP_006252702.1 | 7 - 8 | 853 | 79 | ||
XM_006252641.2 | XP_006252703.1 | 6 - 7 | 994 | 79 | ||
XM_006252644.2 | XP_006252706.1 | 7 - 8 | 853 | 79 | ||
XM_006252645.2 | XP_006252707.1 | 6 - 7 | 994 | 79 | ||
XM_006252647.1 | XP_006252709.1 | 7 - 8 | 853 | 79 | ||
GenBank mRNA | BC083721.1 | - | 6 - 7 | 745 | 7190925 | 79 |
FM122871.1 | - | 5 - 6 | 563 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |