Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100989.1 | NP_001094459.1 | 6 - 7 | 703 | 84 | |
XM_008768993.1 | XP_008767215.1 | 10 - 11 | 1608 | 84 | ||
XM_008768994.1 | XP_008767216.1 | 10 - 11 | 1462 | 84 | ||
GenBank mRNA | BC127538.1 | - | 6 - 7 | 703 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |