Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144759.2 | NP_653360.2 | 2 - 3 | 265 | 104 | |
XM_006248991.2 | XP_006249053.1 | 2 - 3 | 1051 | 104 | ||
GenBank mRNA | CB765131.1 | - | 2 - 3 | 265 | 104 | |
EU741665.1 | - | 2 - 3 | 245 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |