Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769055.1 | XP_008767277.1 | 4 - 5 | 1402 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |