Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013223.1 | NP_037355.1 | 13 - 14 | 1532 | 66 | |
GenBank mRNA | BC081838.1 | - | 13 - 14 | 1565 | 7132671 | 66 |
FQ227635.1 | - | 2 - 3 | 164 | 66 | ||
L27707.1 | - | 13 - 14 | 1532 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |