Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013111.3 | NP_037243.2 | 636 | 68 | ||
XM_008768943.1 | XP_008767165.1 | 1 - 2 | 908 | 68 | ||
XM_008768944.1 | XP_008767166.1 | 1 - 2 | 908 | 68 | ||
GenBank mRNA | AB066224.1 | - | 1 - 2 | 393 | 68 | |
AF245000.1 | - | 576 | 68 | |||
D67087.1 | - | 1 - 2 | 366 | 68 | ||
L10152.1 | - | 1 - 2 | 360 | 68 | ||
U70476.1 | - | 1 - 2 | 445 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |