Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012743.1 | NP_001012761.1 | 4 - 5 | 729 | 98 | |
XM_008760304.1 | XP_008758526.1 | 4 - 5 | 360 | 98 | ||
GenBank mRNA | BC090349.1 | - | 4 - 5 | 729 | 7311399 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |