Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053509.1 | NP_445961.1 | 9 - 10 | 1579 | 63 | |
GenBank mRNA | AB000928.1 | - | 9 - 10 | 1579 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |