Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133535.2 | NP_598219.2 | 12 - 13 | 885 | 76 | |
XM_006231192.2 | XP_006231254.1 | 10 - 11 | 871 | 76 | ||
XM_008760303.1 | XP_008758525.1 | 11 - 12 | 916 | 76 | ||
GenBank mRNA | AF353305.1 | - | 12 - 13 | 941 | 76 | |
BC086376.1 | - | 12 - 13 | 885 | 7135348 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |