Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006972.1 | NP_001006973.1 | 9 - 10 | 1321 | 69 | |
GenBank mRNA | BC083709.1 | - | 9 - 10 | 1321 | 7190583 | 69 |
FQ214688.1 | - | 9 - 10 | 1329 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |