Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134760.1 | NP_001128232.1 | 4 - 5 | 698 | 82 | |
NM_032619.2 | NP_116008.2 | 4 - 5 | 698 | 82 | ||
XM_006228691.2 | XP_006228753.1 | 4 - 5 | 842 | 82 | ||
XM_006228692.2 | XP_006228754.1 | 4 - 5 | 842 | 82 | ||
GenBank mRNA | BC169048.1 | - | 4 - 5 | 653 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |