Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012714.1 | NP_036846.1 | 6 - 7 | 645 | 123 | |
GenBank mRNA | L19660.1 | - | 6 - 7 | 645 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |