Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012859.1 | NP_036991.1 | 3 - 4 | 1698 | 139 | |
XM_006228391.2 | XP_006228453.1 | 3 - 4 | 831 | 139 | ||
XM_006228393.2 | XP_006228455.1 | 3 - 4 | 1082 | 139 | ||
XM_006228394.2 | XP_006228456.1 | 3 - 4 | 2409 | 139 | ||
XM_008758895.1 | XP_008757117.1 | 3 - 4 | 815 | 139 | ||
XM_008758896.1 | XP_008757118.1 | 3 - 4 | 975 | 139 | ||
GenBank mRNA | U40001.1 | - | 3 - 4 | 1698 | 139 | |
X51415.1 | - | 2 - 3 | 1220 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |