Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfnb31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181088.1 | NP_851602.1 | 4 - 5 | 1165 | 113 | |
GenBank mRNA | AY227205.1 | - | 4 - 5 | 1165 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |