Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011976.2 | NP_001011976.2 | 1 - 2 | 60 | 125 | |
XM_006238239.2 | XP_006238301.1 | 1 - 2 | 89 | 125 | ||
GenBank mRNA | BC079286.1 | - | 1 - 2 | 60 | 7134286 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |