Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011976.2 | NP_001011976.2 | 7 - 8 | 764 | 82 | |
XM_006238237.2 | XP_006238299.1 | 7 - 8 | 728 | 82 | ||
XM_006238239.2 | XP_006238301.1 | 7 - 8 | 790 | 82 | ||
GenBank mRNA | BC079286.1 | - | 7 - 8 | 764 | 7134286 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |