Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053288.2 | NP_445740.1 | 5 - 6 | 576 | 80 | |
GenBank mRNA | FQ209739.1 | - | 5 - 6 | 578 | 80 | |
FQ210338.1 | - | 5 - 6 | 578 | 80 | ||
FQ218200.1 | - | 5 - 6 | 581 | 80 | ||
FQ218302.1 | - | 5 - 6 | 578 | 80 | ||
FQ219945.1 | - | 5 - 6 | 582 | 80 | ||
FQ220896.1 | - | 5 - 6 | 582 | 80 | ||
FQ221011.1 | - | 5 - 6 | 581 | 80 | ||
FQ221095.1 | - | 5 - 6 | 577 | 80 | ||
FQ221803.1 | - | 4 - 5 | 372 | 80 | ||
FQ221848.1 | - | 5 - 6 | 578 | 80 | ||
FQ222215.1 | - | 5 - 6 | 578 | 80 | ||
FQ222231.1 | - | 4 - 5 | 389 | 80 | ||
FQ222829.1 | - | 5 - 6 | 578 | 80 | ||
FQ223035.1 | - | 5 - 6 | 578 | 80 | ||
FQ228353.1 | - | 5 - 6 | 492 | 80 | ||
FQ228371.1 | - | 5 - 6 | 582 | 80 | ||
FQ228464.1 | - | 5 - 6 | 576 | 80 | ||
FQ228489.1 | - | 5 - 6 | 578 | 80 | ||
FQ228584.1 | - | 5 - 6 | 578 | 80 | ||
FQ228800.1 | - | 5 - 6 | 578 | 80 | ||
FQ229026.1 | - | 5 - 6 | 577 | 80 | ||
FQ229053.1 | - | 4 - 5 | 302 | 80 | ||
J00696.1 | - | 5 - 6 | 577 | 80 | ||
V01216.1 | - | 5 - 6 | 576 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |