Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232783.2 | XP_006232845.1 | 15 - 16 | 2744 | 121 | |
XM_006232784.2 | XP_006232846.1 | 16 - 17 | 3089 | 121 | ||
XM_006232785.2 | XP_006232847.1 | 13 - 14 | 2536 | 121 | ||
XM_215597.8 | XP_215597.6 | 15 - 16 | 2742 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |