Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232783.2 | XP_006232845.1 | 2 - 3 | 410 | 129 | |
XM_006232784.2 | XP_006232846.1 | 2 - 3 | 715 | 169 | ||
XM_006232785.2 | XP_006232847.1 | 2 - 3 | 412 | 129 | ||
XM_215597.8 | XP_215597.6 | 2 - 3 | 408 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |