Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107702.1 | NP_001101172.1 | 2 - 3 | 436 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |